Merck
Identification of marker genes for spinal cord injury.
Frontiers in medicine (2024-03-11)
Zhiwei Luan, Jiayu Zhang, Yansong Wang
ABSTRACT
Spinal cord injury (SCI) is a profoundly disabling and devastating neurological condition, significantly impacting patients' quality of life. It imposes unbearable psychological and economic pressure on both patients and their families, as well as placing a heavy burden on society. In this study, we integrated datasets GSE5296 and GSE47681 as training groups, analyzed gene variances between sham group and SCI group mice, and conducted Gene Ontology (GO) enrichment analysis and Kyoto Encyclopedia of Genes and Genomes (KEGG) enrichment analysis based on the differentially expressed genes. Subsequently, we performed Weighted Gene Correlation Network Analysis (WGCNA) and Lasso regression analyses. We identified four characteristic disease genes: Icam1, Ch25h, Plaur and Tm4sf1. We examined the relationship between SCI and immune cells, and validated the expression of the identified disease-related genes in SCI rats using PCR and immunohistochemistry experiments. In conclusion, we have identified and verified four genes related to SCI: Icam1, Ch25h, Plaur and Tm4sf1, which could offer insights for SCI treatment.