Merck
  • Convergent activation of Ca2+ permeability in two-pore channel 2 through distinct molecular routes.
Convergent activation of Ca2+ permeability in two-pore channel 2 through distinct molecular routes.
Science signaling (2023-08-22)
Ryo Saito, Qianru Mu, Yu Yuan, Marcos Rubio-Alarcón, Maria Eznarriaga, Pingwei Zhao, Gihan Gunaratne, Sushil Kumar, Marco Keller, Franz Bracher, Christian Grimm, Eugen Brailoiu, Jonathan S Marchant, Taufiq Rahman, Sandip Patel
ABSTRACT
TPC2 is a pathophysiologically relevant lysosomal ion channel that is activated directly by the phosphoinositide PI(3,5)P2 and indirectly by the calcium ion (Ca2+)-mobilizing molecule NAADP through accessory proteins that associate with the channel. TPC2 toggles between PI(3,5)P2-induced, sodium ion (Na+)-selective and NAADP-induced, Ca2+-permeable states in response to these cues. To address the molecular basis of polymodal gating and ion-selectivity switching, we investigated the mechanism by which NAADP and its synthetic functional agonist, TPC2-A1-N, induced Ca2+ release through TPC2 in human cells. Whereas NAADP required the NAADP-binding proteins JPT2 and LSM12 to evoke endogenous calcium ion signals, TPC2-A1-N did not. Residues in TPC2 that bind to PI(3,5)P2 were required for channel activation by NAADP but not for activation by TPC2-A1-N. The cryptic voltage-sensing region of TPC2 was required for the actions of TPC2-A1-N and PI(3,5)P2 but not for those of NAADP. These data mechanistically distinguish natural and synthetic agonist action at TPC2 despite convergent effects on Ca2+ permeability and delineate a route for pharmacologically correcting impaired NAADP-evoked Ca2+ signals.