Merck
  • TLR2 Supports γδ T cell IL-17A Response to ocular surface commensals by Metabolic Reprogramming.
TLR2 Supports γδ T cell IL-17A Response to ocular surface commensals by Metabolic Reprogramming.
bioRxiv : the preprint server for biology (2024-05-07)
Wenjie Zhu, Xiaoyan Xu, Vijayaraj Nagarajan, Jing Guo, Zixuan Peng, Amy Zhang, Jie Liu, Mary J Mattapallil, Yingyos Jittayasothorn, Reiko Horai, Anthony J St Leger, Rachel R Caspi
ABSTRACT
The ocular surface is a mucosal barrier tissue colonized by commensal microbes, which tune local immunity by eliciting IL-17 from conjunctival γδ T cells to prevent pathogenic infection. The commensal Corynebacterium mastitidis (C. mast) elicits protective IL-17 responses from conjunctival Vγ4 T cells through a combination of γδ TCR ligation and IL-1 signaling. Here, we identify Vγ6 T cells as a major C. mast-responsive subset in the conjunctiva and uncover its unique activation requirements. We demonstrate that Vγ6 cells require not only extrinsic (via dendritic cells) but also intrinsic TLR2 stimulation for optimal IL-17A response. Mechanistically, intrinsic TLR2 signaling was associated with epigenetic changes and enhanced expression of genes responsible for metabolic shift to fatty acid oxidation to support Il17a transcription. We identify one key transcription factor, IκBζ, which is upregulated by TLR2 stimulation and is essential for this program. Our study highlights the importance of intrinsic TLR2 signaling in driving metabolic reprogramming and production of IL-17A in microbiome-specific mucosal γδ T cells.