Merck
  • The hippocampal FTO-BDNF-TrkB pathway is required for novel object recognition memory reconsolidation in mice.
The hippocampal FTO-BDNF-TrkB pathway is required for novel object recognition memory reconsolidation in mice.
Translational psychiatry (2023-11-15)
Rui Chang, Shanshan Zhu, Jionghong Peng, Zhenyi Lang, Xinyu Zhou, Hailin Liao, Ju Zou, Peng Zeng, Sijie Tan
ABSTRACT
Memory reconsolidation refers to the process by which the consolidated memory was restored after reactivation (RA). Memory trace becomes labile after reactivation and inhibition of memory reconsolidation may disrupt or update the original memory trace, which provided a new strategy for the treatment of several psychiatric diseases, such as drug addiction and post-traumatic stress disorder. Fat mass and obesity-associated gene (FTO) is a novel demethylase of N6-methyladenosine (m6A) and it has been intensively involved in learning and memory. However, the role of FTO in memory reconsolidation has not been determined. In the present study, the function of FTO in memory reconsolidation was investigated in the novel object recognition (NOR) model in mice. The results showed that RA of NOR memory increased hippocampal FTO expression in a time-dependent manner, while FTO inhibitor meclofenamic acid (MA) injected immediately, but not 6 h after RA disrupted NOR memory reconsolidation. MA downregulated BDNF expression during NOR memory reconsolidation in the hippocampus, while the TrkB agonist 7,8-Dihydroxyflavone (7,8-DHF) reversed the disruptive effects of MA on NOR memory reconsolidation. Furthermore, overexpression of FTO increased BDNF expression via decreasing mRNA m6A in HT22 cells. Taken together, these results indicate that FTO may up-regulate the BDNF-TrkB pathway to promote NOR memory reconsolidation through m6A modification.