Merck
  • Schwann cell-derived amphiregulin enhances nerve regeneration via supporting the proliferation and migration of Schwann cells and the elongation of axons.
Schwann cell-derived amphiregulin enhances nerve regeneration via supporting the proliferation and migration of Schwann cells and the elongation of axons.
Journal of neurochemistry (2023-07-13)
Sailing Chen, Qianqian Chen, Xiaojiao Zhang, Yinying Shen, Xinyu Shi, Xiu Dai, Sheng Yi
ABSTRACT
Peripheral nerves have limited regeneration ability following nerve injury. Applying growth factors with neurotrophic roles is beneficial for accelerating peripheral nerve regeneration. Here we show that after rat sciatic nerve injury, growth factor amphiregulin (AREG) is upregulated in Schwann cells of sciatic nerves. Elevated AREG stimulates the proliferation and migration of Schwann cells by activating ERK1/2 cascade. Schwann cell-secreted AREG further facilitates the outgrowth of neurites and the elongation of injured axons. Administration of AREG to injured sciatic nerves stimulates the proliferation of Schwann cells to replace lost cell population, encourages the migration of Schwann cells to form cell cords, and facilitates the regrowth of axons. Overall, our results identify AREG as an important neurotrophic factor and thus provide a promising therapeutic avenue towards peripheral nerve injury.
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder