Merck
  • Exacerbation of Cisplatin Cellular Toxicity by Regulation of the Human Organic Cation Transporter 2 through Angiotensin II.
Exacerbation of Cisplatin Cellular Toxicity by Regulation of the Human Organic Cation Transporter 2 through Angiotensin II.
International journal of molecular sciences (2022-12-24)
Marta Kantauskaite, Anna Hucke, Beatrice Snieder, Giuliano Ciarimboli
ABSTRACT
Cisplatin (CDDP) is an efficient chemotherapeutic drug, whose use is associated with the development of serious undesired toxicities, such as nephrotoxicity. The human organic cation transporter 2 (hOCT2), which is highly expressed in the basolateral membrane domain of renal proximal tubules seems to play an important role in the development of CDDP nephrotoxicity. The role of angiotensin II (AII) signaling by binding to the AII receptor type 1 (AT1R) in the development and/or progression of CDDP nephrotoxicity is debated. Therefore, in this work, the regulation of hOCT2 activity by AII and its role in the development of CDDP cellular toxicity was investigated. To do this, hOCT2 was overexpressed by viral transduction in Madin-Darby Canine Kidney (MDCK) cells which were cultivated on a filter. This approach allows the separation of an apical and a basolateral membrane domain, which are easily accessible for experimentation. In this system, hOCT2 was mainly localized on the basolateral plasma membrane domain of the cells. The transporter was functional since a specific uptake of the fluorescent organic cation 4-(4-(dimethylamino)styryl)-N-methylpyridinium (ASP+) with an affinity (Km) of 35 µM was only detectable by the addition of ASP+ to the basolateral compartment of hOCT2 expressing MDCK (hOCT2-MDCK) cells. Similarly, CDDP toxicity was evident mainly by CDDP addition to the basolateral compartment of hOCT2-MDCK cells cultivated on a filter. The addition of 1 nM AII stimulated hOCT2 function via PKC activation and worsened CDDP cytotoxicity via binding to AT1R. Therefore, the AII signaling pathway may be implicated in the development and/or progression of CDDP nephrotoxicity. This signaling pathway may be a target for protective interventions for example by blocking AT1R in the kidneys. However, it should be further investigated whether these findings obtained in a cell culture system may have translational relevance for the clinical situation. For toxicity experiments, a 100 µM CDDP concentration was used, which is high but allows us to identify clearly toxic effects due to hOCT2. In summary, down-regulation of hOCT2 activity by the inhibition of the AII signaling pathway may protect against CDDP nephrotoxicity.