Merck
  • Isolation and Characterization of Poeciguamerin, a Peptide with Dual Analgesic and Anti-Thrombotic Activity from the Poecilobdella manillensis Leech.
Isolation and Characterization of Poeciguamerin, a Peptide with Dual Analgesic and Anti-Thrombotic Activity from the Poecilobdella manillensis Leech.
International journal of molecular sciences (2023-07-14)
Chaoming Wang, Mengrou Chen, Xiaoyu Lu, Shuo Yang, Min Yang, Yaqun Fang, Ren Lai, Zilei Duan
ABSTRACT
When Poecilobdella manillensis attacks its prey, the prey bleeds profusely but feels little pain. We and other research teams have identified several anticoagulant molecules in the saliva of P. manillensis, but the substance that produces the paralyzing effect in P. manillensis is not known. In this study, we successfully isolated, purified, and identified a serine protease inhibitor containing an antistasin-like domain from the salivary secretions of P. manillensis. This peptide (named poeciguamerin) significantly inhibited elastase activity and slightly inhibited FXIIa and kallikrein activity, but had no effect on FXa, trypsin, or thrombin activity. Furthermore, poeciguamerin exhibited analgesic activity in the foot-licking and tail-withdrawal mouse models and anticoagulant activity in the FeCl3-induced carotid artery thrombosis mouse model. In this study, poeciguamerin was found to be a promising elastase inhibitor with potent analgesic and antithrombotic activity for the inhibition of pain and thrombosis after surgery or in inflammatory conditions.
Product Description
Trypsin, BRP, European Pharmacopoeia (EP) Reference Standard