Merck
  • In Vitro Models to Evaluate Molecular Permeability of the Kidney Filtration Barrier.
In Vitro Models to Evaluate Molecular Permeability of the Kidney Filtration Barrier.
Methods in molecular biology (Clifton, N.J.) (2023-07-10)
Dan Wang, Nicholas Ferrell
ABSTRACT
The glomerular basement membrane (GBM) is an important component of the kidney filtration barrier. The ability to evaluate the molecular transport properties of the GBM and determining how changes in the structure, composition, and mechanical properties of the GBM regulate its size selective transport properties may provide additional insight into glomerular function. This chapter details a method for making in vitro models of the glomerular filtration barrier using animal-derived decellularized glomeruli. FITC-labelled Ficoll is used as a filtration probe to evaluate the molecular transport properties during passive diffusion and under applied pressure. These systems can serve as a platform to evaluate the molecular permeability of basement membrane systems using conditions that simulate normal or pathophysiological conditions.