Merck
  • Fibroblast growth factor 10 protects against UVB-induced skin injury by activating the ERK/YAP signalling pathway.
Fibroblast growth factor 10 protects against UVB-induced skin injury by activating the ERK/YAP signalling pathway.
Cell proliferation (2022-07-20)
Nan Wang, Yetong Dong, Xiejun Xu, Yingjie Shen, Zhiyuan Huang, Yin Yu, Zhili Liu, Wenjie Gong, Siyi Zhang, Yeyi Zheng, Yonghuan Song, Zhongxin Zhu, Litai Jin, Weitao Cong
ABSTRACT
Ultraviolet light B (UVB) irradiation can induce skin injury and result in keratinocytes proliferation inhibition. However, the molecular understanding of the repair during UVB-induced cell proliferation inhibition remains poorly understood. The purpose of this study was to explore the role and potential mechanism of FGF10 in promoting keratinocytes cell cycle and proliferation after UVB injury. Expression of FGF10 protein was analysed in skin treated with UVB radiation by immunohistochemistry. The proliferation potential was examined by Immunofluorescence, Western Blot and RT-PCR under UVB radiation, treated with FGF10 protein or overexpression of FGF10 using adeno-associated virus. CCK8 kit was used to further detect cell proliferation ability. We found that FGF10 is highly expressed in skin treated with UVB. Overexpression of FGF10 has a protective effect against UVB-induced skin damage by balancing epidermal thickness and enhancing epidermal keratinocytes proliferation. Importantly, FGF10 is found to alleviate UVB-induced downregulation of YAP activity, then promoting keratinocytes proliferation. Disruption of YAP function, either with the small molecule YAP inhibitor Verteporfin (VP) or YAP small-interfering RNA (siRNA), largely abolishes the protective activity of FGF10 on epidermal keratinocytes proliferation. Meanwhile, disruption of ERK kinase (MEK) activity with U0126 or ERK siRNA hinder the positive influence of FGF10 on UVB-induced skin injury. FGF10 promotes epidermal keratinocytes proliferation during UVB-induced skin injury in an ERK/YAP-dependent manner.