Merck
  • Serological Investigation of Bovine Toxoplasmosis Using Commercial and Indigenous ELISA Kits While Validating Cattle Toxo IgG ELISA Kit.
Serological Investigation of Bovine Toxoplasmosis Using Commercial and Indigenous ELISA Kits While Validating Cattle Toxo IgG ELISA Kit.
Animals : an open access journal from MDPI (2022-08-27)
Sarfraz-Ur-Rahman, Haroon Akbar, Muhammad Zubair Shabbir, Ubaid Ullah, Muhammad Imran Rashid
ABSTRACT
Toxoplasma gondii is a protozoan parasite that causes toxoplasmosis in warm-blooded vertebrates, globally. The main aims of this study were to assess the seropositivity to toxoplasmosis of an exotic breed of cattle (n = 400) from different farms using the Latex Agglutination Test and validate Cattle Toxo IgG ELISA kit. Of a total of 400 cattle sera that were evaluated by LAT, 143 (35.75%) were found positive. Based on these data, 90 samples (n = 60 seronegative by LAT; n = 30 seropositive by LAT) were elected for screening through a commercially available ELISA kit. The same 90 samples were screened through a Cattle Toxo IgG ELISA kit for validation purposes. Of 90 samples, 40 were seropositive in the Cattle Toxo IgG ELISA kit (100% sensitivity), and 38 were seropositive in a commercially available ELISA kit. All 50 samples in the Cattle Toxo IgG ELISA kit (96.15% specificity) were also seronegative in the commercially available ELISA kit. Hence, the sensitivity and specificity of the Cattle Toxo IgG ELISA kit came out to be 100% and 96.15%, and in LAT, it was found as 26.31% and 61.53%, respectively. Therefore, the Cattle Toxo IgG ELISA kit is a highly reliable serodiagnostic tool to diagnose bovine toxoplasmosis.