Merck
Mast cell chymase has a negative impact on human osteoblasts.
Matrix biology : journal of the International Society for Matrix Biology (2022-08-01)
Thomas Lind, Fabio Rabelo Melo, Ann-Marie Gustafson, Anders Sundqvist, Xinran O Zhao, Aristidis Moustakas, Håkan Melhus, Gunnar Pejler
ABSTRACT
Mast cells have been linked to osteoporosis and bone fractures, and in a previous study we found that mice lacking a major mast cell protease, chymase, develop increased diaphyseal bone mass. These findings introduce the possibility that mast cell chymase can regulate bone formation, but the underlying mechanism(s) has not previously been investigated. Here we hypothesized that chymase might exert such effects through a direct negative impact on osteoblasts, i.e., the main bone-building cells. Indeed, we show that chymase has a distinct impact on human primary osteoblasts. Firstly, chymase was shown to have pronounced effects on the morphological features of osteoblasts, including extensive cell contraction and actin reorganization. Chymase also caused a profound reduction in the output of collagen from the osteoblasts, and was shown to degrade osteoblast-secreted fibronectin and to activate pro-matrix metallopeptidase-2 released by the osteoblasts. Further, chymase was shown to have a preferential impact on the gene expression, protein output and phosphorylation status of TGFβ-associated signaling molecules. A transcriptomic analysis was conducted and revealed a significant effect of chymase on several genes of importance for bone metabolism, including a reduction in the expression of osteoprotegerin, which was confirmed at the protein level. Finally, we show that chymase interacts with human osteoblasts and is taken up by the cells. Altogether, the present findings provide a functional link between mast cell chymase and osteoblast function, and can form the basis for a further evaluation of chymase as a potential target for intervention in metabolic bone diseases.