Merck
  • Rotary properties of hybrid F1-ATPases consisting of subunits from different species.
Rotary properties of hybrid F1-ATPases consisting of subunits from different species.
iScience (2023-05-17)
Ryo R Watanabe, Busra Tas Kiper, Mariel Zarco-Zavala, Mayu Hara, Ryohei Kobayashi, Hiroshi Ueno, José J García-Trejo, Chun-Biu Li, Hiroyuki Noji
ABSTRACT
F1-ATPase (F1) is an ATP-driven rotary motor protein ubiquitously found in many species as the catalytic portion of FoF1-ATP synthase. Despite the highly conserved amino acid sequence of the catalytic core subunits: α and β, F1 shows diversity in the maximum catalytic turnover rate Vmax and the number of rotary steps per turn. To study the design principle of F1, we prepared eight hybrid F1s composed of subunits from two of three genuine F1s: thermophilic Bacillus PS3 (TF1), bovine mitochondria (bMF1), and Paracoccus denitrificans (PdF1), differing in the Vmax and the number of rotary steps. The Vmax of the hybrids can be well fitted by a quadratic model highlighting the dominant roles of β and the couplings between α-β. Although there exist no simple rules on which subunit dominantly determines the number of steps, our findings show that the stepping behavior is characterized by the combination of all subunits.