Merck
  • Human immunoglobulin repertoire analysis guides design of vaccine priming immunogens targeting HIV V2-apex broadly neutralizing antibody precursors.
Human immunoglobulin repertoire analysis guides design of vaccine priming immunogens targeting HIV V2-apex broadly neutralizing antibody precursors.
Immunity (2022-10-01)
Jordan R Willis, Zachary T Berndsen, Krystal M Ma, Jon M Steichen, Torben Schiffner, Elise Landais, Alessia Liguori, Oleksandr Kalyuzhniy, Joel D Allen, Sabyasachi Baboo, Oluwarotimi Omorodion, Jolene K Diedrich, Xiaozhen Hu, Erik Georgeson, Nicole Phelps, Saman Eskandarzadeh, Bettina Groschel, Michael Kubitz, Yumiko Adachi, Tina-Marie Mullin, Nushin B Alavi, Samantha Falcone, Sunny Himansu, Andrea Carfi, Ian A Wilson, John R Yates, James C Paulson, Max Crispin, Andrew B Ward, William R Schief
ABSTRACT
Broadly neutralizing antibodies (bnAbs) to the HIV envelope (Env) V2-apex region are important leads for HIV vaccine design. Most V2-apex bnAbs engage Env with an uncommonly long heavy-chain complementarity-determining region 3 (HCDR3), suggesting that the rarity of bnAb precursors poses a challenge for vaccine priming. We created precursor sequence definitions for V2-apex HCDR3-dependent bnAbs and searched for related precursors in human antibody heavy-chain ultradeep sequencing data from 14 HIV-unexposed donors. We found potential precursors in a majority of donors for only two long-HCDR3 V2-apex bnAbs, PCT64 and PG9, identifying these bnAbs as priority vaccine targets. We then engineered ApexGT Env trimers that bound inferred germlines for PCT64 and PG9 and had higher affinities for bnAbs, determined cryo-EM structures of ApexGT trimers complexed with inferred-germline and bnAb forms of PCT64 and PG9, and developed an mRNA-encoded cell-surface ApexGT trimer. These methods and immunogens have promise to assist HIV vaccine development.