Merck
  • Accumulated cholesterol protects tumours from elevated lipid peroxidation in the microenvironment.
Accumulated cholesterol protects tumours from elevated lipid peroxidation in the microenvironment.
Redox biology (2023-03-21)
Xi Zhao, Xinyu Lian, Jianlan Xie, Guoquan Liu
ABSTRACT
Elevated lipid peroxidation (LPO), usually present in the tumour microenvironment (TME), is profoundly implicated in antitumour immunity and may be targeted for the development of new antitumour therapies. However, tumour cells may also rewire their metabolism to survive elevated LPO. Here, we report a novel and nonantioxidant mechanism by which tumour cells benefit from accumulated cholesterol to restrain LPO and ferroptosis, a nonapoptotic form of cell death characterized by accumulated LPO. Modulating cholesterol metabolism, especially LDLR-mediated cholesterol uptake, shifted the susceptibility of tumour cells to ferroptosis. Elevation of cellular cholesterol content specifically restrained LPO triggered by GSH-GPX4 inhibition or oxidizing factors in the TME. Furthermore, depletion of TME cholesterol by MβCD efficiently enhanced the antitumour efficacy of ferroptosis in a mouse xenograft model. Distinct from the antioxidant effect of its metabolic intermediates, the protective role of cholesterol was ascribed to its ability to decrease membrane fluidity and promote lipid raft formation, which affects the diffusion of LPO substrates. A correlation between LPO and lipid rafts was also found in tumour tissues from renal cancer patients. Together, our findings have identified a general and nonsacrificial mechanism by which cholesterol suppresses LPO, which can be exploited to enhance the efficacy of ferroptosis-based antitumour strategies.