Merck
Two Distant Catalytic Sites Are Responsible for C2c2 RNase Activities.
Cell (2017-01-14)
Liang Liu, Xueyan Li, Jiuyu Wang, Min Wang, Peng Chen, Maolu Yin, Jiazhi Li, Gang Sheng, Yanli Wang
ABSTRACT
C2c2, the effector of type VI CRISPR-Cas systems, has two RNase activities-one for cutting its RNA target and the other for processing the CRISPR RNA (crRNA). Here, we report the structures of Leptotrichia shahii C2c2 in its crRNA-free and crRNA-bound states. While C2c2 has a bilobed structure reminiscent of all other Class 2 effectors, it also exhibits different structural characteristics. It contains the REC lobe with a Helical-1 domain and the NUC lobe with two HEPN domains. The two RNase catalytic pockets responsible for cleaving pre-crRNA and target RNA are independently located on Helical-1 and HEPN domains, respectively. crRNA binding induces significant conformational changes that are likely to stabilize crRNA binding and facilitate target RNA recognition. These structures provide important insights into the molecular mechanism of dual RNase activities of C2c2 and establish a framework for its future engineering as a RNA editing tool.