- Photopharmacological control of cell signaling with photoswitchable lipids.
Photopharmacological control of cell signaling with photoswitchable lipids.
Current opinion in pharmacology (2022-03-13)
Tufan K Mukhopadhyay, Johannes Morstein, Dirk Trauner
PMID35278838
ABSTRACT
Many aspects of cell signaling are controlled by lipids. Several signaling lipids have been functionalized with an azobenzene photoswitch to control underlying signaling dynamics with light. Herein, we provide an overview of signaling photolipids developed to date focusing on their biological applications. These include immunomodulation, kinase translocation, apoptosis, ion channel activation, G-protein coupled receptor activation, and neurogenesis.