- Extracellular superoxide dismutase and cardiovascular disease.
Extracellular superoxide dismutase and cardiovascular disease.
Cardiovascular research (2002-07-19)
Tohru Fukai, Rodney J Folz, Ulf Landmesser, David G Harrison
PMID12123763
ABSTRACT
Excessive production and/or inadequate removal of reactive oxygen species, especially superoxide anion (O(2)(*-)), have been implicated in the pathogenesis of many cardiovascular diseases, including atherosclerosis, hypertension, diabetes, and in endothelial dysfunction by decreasing nitric oxide (NO) bioactivity. Since the vascular levels of O(2)(*-) are regulated by the superoxide dismutase (SOD) enzymes, a role of SOD in the cardiovascular disease is of substantial interest. Particularly, a major form of SOD in the vessel wall is the extracellular SOD (ecSOD). This review will discuss the characteristics of ecSOD and the role of ecSOD in cardiovascular diseases.