- Analysis of Golgi Secretory Functions in Cancer.
Analysis of Golgi Secretory Functions in Cancer.
Methods in molecular biology (Clifton, N.J.) (2022-12-14)
Priyam Banerjee, Xiaochao Tan, William K Russell, Jonathan M Kurie
PMID36512251
ABSTRACT
Cancer cells utilize secretory pathways for paracrine signaling and extracellular matrix remodeling to facilitate directional cell migration, invasion, and metastasis. The Golgi apparatus is a central secretory signaling hub that is often deregulated in cancer. Here we described technologies that utilize microscopic, biochemical, and proteomic approaches to analyze Golgi secretory functions in genetically heterogeneous cancer cell lines.