Merck
ProNGF Expression and Targeting in Glioblastoma Multiforme.
International journal of molecular sciences (2023-01-22)
Mark Marsland, Amiee Dowdell, Sam Faulkner, Phillip Jobling, Robert A Rush, Craig Gedye, James Lynam, Cassandra P Griffin, Mark Baker, Joanne Marsland, Chen Chen Jiang, Hubert Hondermarck
ABSTRACT
Glioblastoma multiforme (GBM) is the most lethal adult brain cancer. Temozolomide (TMZ), the standard chemotherapeutic drug used in GBM, has limited benefit and alternate therapies are needed to improve GBM treatment. Nerve growth factor (NGF) and its precursor proNGF are increasingly recognized as stimulators of human tumor progression. The expression and stimulatory effect of NGF on GBM cell growth has previously been reported, but the status of proNGF in GBM is unreported. In this study, we have investigated proNGF expression and biological activity in GBM. A clinical cohort of GBM (n = 72) and low-grade glioma (n = 20) was analyzed by immunohistochemistry for proNGF and digital quantification. ProNGF expression was significantly increased in GBM compared to low grade gliomas and proNGF was also detected in patient plasma samples. ProNGF was also detected in most GBM cell lines by Western blotting. Although anti-proNGF blocking antibodies inhibited cell growth in GBM cells with methylated MGMT gene promoter, targeting proNGF could not potentiate the efficacy of TMZ. In subcutaneous xenograft of human GBM cells, anti-proNGF antibodies slightly reduced tumor volume but had no impact on TMZ efficacy. In conclusion, this data reveals that proNGF is overexpressed in GBM and can stimulate cancer cell growth. The potential of proNGF as a clinical biomarker and therapeutic target warrants further investigations.
Product Description
Corning® 96 Well TC-Treated Microplates, size 96 wells, polystyrene, flat bottom, case of 100 (individually wrapped), sterile, lid