Merck
  • Saccharopolyspora Species: Laboratory Maintenance and Enhanced Production of Secondary Metabolites.
Saccharopolyspora Species: Laboratory Maintenance and Enhanced Production of Secondary Metabolites.
Current protocols in microbiology (2017-02-07)
Dipesh Dhakal, Anaya Raj Pokhrel, Amit Kumar Jha, Nguyen Huy Thuan, Jae Kyung Sohng
ABSTRACT
Saccharopolyspora spp. are aerobic, Gram-positive, non-acid-fast, and non-motile actinomycetes. Various species of the genus Saccharopolyspora have been reported with an ability to produce various bioactive compounds for pharmaceutical and agricultural uses. This unit includes general protocols for the laboratory maintenance of Saccharopolyspora species, including growth in liquid medium, growth on solid agar, long-term storage, and generation of a higher producer strain by mutagenesis. Saccharopolyspora spinosa ATCC 49460 is used as a prototype for explaining the considerations for efficient laboratory maintenance of Saccharopolyspora spp. Saccharopolyspora spinosa is a producer of spinosad, a prominent insecticide with selective activity against various insects. © 2017 by John Wiley & Sons, Inc.