Merck
  • The ubiquitin ligase c-Cbl down-regulates FcgammaRIIa activation in human neutrophils.
The ubiquitin ligase c-Cbl down-regulates FcgammaRIIa activation in human neutrophils.
Journal of immunology (Baltimore, Md. : 1950) (2009-02-10)
Louis Marois, Myriam Vaillancourt, Sébastien Marois, Sophie Proulx, Guillaume Paré, Emmanuelle Rollet-Labelle, Paul H Naccache
ABSTRACT
Little is known about the mechanisms that arrest FcgammaRIIa signaling in human neutrophils once engaged by immune complexes or opsonized pathogens. In our previous studies, we observed a loss of immunoreactivity of Abs directed against FcgammaRIIa following its cross-linking. In this study, we report on the mechanisms involved in this event. A stimulated internalization of FcgammaRIIa leading to the down-regulation of its surface expression was observed by flow cytometry and confocal microscopy. Immunoprecipitation of the receptor showed that FcgammaRIIa is ubiquitinated after stimulation. MG132 and clasto-lactacystin beta-lactone inhibited the loss of immunoreactivity of FcgammaRIIa, suggesting that this receptor was down-regulated via the proteasomal pathway. The E3 ubiquitin ligase c-Cbl was found to translocate from the cytosol to the plasma membrane following receptor cross-linking. Furthermore, c-Cbl was recruited to the same subset of high-density, detergent-resistant membrane fractions as stimulated FcgammaRIIa itself. Silencing the expression of c-Cbl by small interfering RNA decreased FcgammaRIIa ubiquitination and prevented its degradation without affecting the internalisation process. It also prolonged the stimulation of the tyrosine phosphorylation response to the cross-linking of the receptor. We conclude that c-Cbl mediates the ubiquitination of stimulated FcgammaRIIa and thereby contributes to the termination of FcgammaRIIa signaling via its proteasomal degradation, thus leading to the down-regulation of neutrophil signalisation and function (phagocytosis) through this receptor.