- Studying the subcellular localization and DNA-binding activity of FoxO transcription factors, downstream effectors of PI3K/Akt.
Studying the subcellular localization and DNA-binding activity of FoxO transcription factors, downstream effectors of PI3K/Akt.
Methods in molecular biology (Clifton, N.J.) (2009-01-24)
Abdelkader Essafi, Ana R Gomes, Karen M Pomeranz, Aleksandra K Zwolinska, Rana Varshochi, Ursula B McGovern, Eric W-F Lam
PMID19160671
ABSTRACT
This chapter describes methods for studying downstream events of the PI3K/Akt signaling cascade, focusing on the FoxO transcription factors. These approaches also represent alternative means for gauging the phosphoinositide-3 kinase/Akt activity. We describe protocols for the fractionation of cytoplasmic and nuclear protein extracts and for studying transcription factor DNA-binding activity in vitro and in vivo.