Merck
  • Regulation of dopaminergic neuronal phenotypes by the estrogen-related receptor gamma ligand GSK4716 via the activation of CREB signaling.
Regulation of dopaminergic neuronal phenotypes by the estrogen-related receptor gamma ligand GSK4716 via the activation of CREB signaling.
Neurochemistry international (2020-03-17)
Juhee Lim, Hyo In Kim, Hyun Jin Choi
ABSTRACT
Midbrain dopaminergic (DAergic) neurotransmission plays a crucial role in regulating motor, cognitive, and emotional functions. The orphan nuclear receptor estrogen-related receptor gamma (ERRγ) is highly expressed in the adult brain and in the developing fetal brain. Our previous study showed the relevance of ERRγ in the regulation of the DAergic neuronal phenotype with the upregulation of dopamine synthesizing tyrosine hydroxylase (TH) and dopamine transporter (DAT) and the possibility that ERRγ could be a novel target for regulating DAergic neuronal differentiation. In this study, we examined whether ERRγ ligands could be small molecule regulators of DAergic phenotypes. The ERRγ agonist GSK4716 increased DAT and TH expression, and the ERRγ inverse agonist GSK5182 attenuated the retinoic acid-induced upregulation of DAT and TH in differentiated SH-SY5Y cells. We found that biphasic activation of the protein kinase A/cyclic AMP response element-binding (CREB) protein signaling pathway was involved in the GSK4716-induced increase in the DAergic phenotype in SH-SY5Y cells. CREB signaling activated as early as 3 h after GSK4716 treatment in an ERRγ-independent manner, but increased following ERRγ activation after 3 days. Protein kinase A inhibitor H-89 attenuated GSK4716-induced DAT and TH upregulation. In primary cultured DAergic neurons, GSK4716 increased neurite length and the number of DAT and TH-double-positive (DAT + TH+) neurons compared to that in control cells. These findings suggest that ERRγ ligands could serve as useful chemical tools for obtaining a better understanding of the regulation of DAergic phenotypes and might facilitate the development of small molecule therapeutics to treat DA-related neurological diseases.