Merck
  • MicroRNA-519d-3p antagonizes osteosarcoma resistance against cisplatin by targeting PD-L1.
MicroRNA-519d-3p antagonizes osteosarcoma resistance against cisplatin by targeting PD-L1.
Molecular carcinogenesis (2021-11-16)
Jing Wang, Zhenjun Zhang, Chuang Qiu, Jiashi Wang
ABSTRACT
Accumulating evidence indicates that a ligand of programmed cell death receptor-1 (PD-L1) participates in the progression and recurrence of multiple malignancies, including osteosarcoma. Nevertheless, the role of PD-L1 in chemoresistance development is not fully understood. In the current study, we aim to clarify the interaction of miR-519d-3p and PD-L1 in the development of cisplatin resistance. Immunohistochemistry, quantitative reverse-transcription polymerase reaction, and Western blot were used to evaluate PD-L1 expression. MTT and transwell migration assays were used to measure cell growth and motility, respectively. ENCORI, miRCode, and miRDB databases were recruited to predict candidate miRNAs targeting PD-L1. The binding sequences of miR-519d-3p and PD-L1 3' untranslated region were identified by dual-luciferase reporter and RNA immunoprecipitation assays. Flow cytometric analysis was conducted to measure the cycle distribution and cell apoptosis. Metastatic mouse models were generated with cisplatin-resistant sublines by intravenous injection. We found that PD-L1 expression was positively correlated to cisplatin resistance and metastasis, whereas miR-519d-3p expression was reduced in cisplatin-resistant specimens and was negatively correlated to cisplatin resistance and metastasis of osteosarcoma. We demonstrated that miR-519d-3p overexpression reversed cisplatin resistance, induced G1/S phase arrest and apoptosis. In addition, we proved that miR-519d-3p inhibited lung metastasis by establishing cisplatin-resistant MG63 metastatic xenograft models. The present findings suggest that miR-519d-3p/PD-L1 axis is a novel signaling pathway contributing to cisplatin resistance. Our study provides new clues for curing refractory osteosarcoma beyond immune checkpoint inhibitors.