Merck
Plasminogen regulates pro-opiomelanocortin processing.
Journal of thrombosis and haemostasis : JTH (2004-04-22)
N Wang, L Zhang, L Miles, J Hoover-Plow
ABSTRACT
Plasminogen-deficient mice exhibit behavioral differences in response to stress, including a markedly reduced acoustic startle reflex response compared with wild-type (WT) littermates. The acoustic startle reflex activates the hypothalamic-pituitary axis and is modulated by these hormones. The purpose of this study was to investigate whether plasminogen plays a role in the processing of hormones in the hypothalamic-pituitary axis. In this study the concentration of plasma, pituitary, and brain hypothalamic-pituitary axis hormones and precursor processing was examined in WT and plasminogen deficient (Plg-/-) mice before and after acoustic startle reflex testing. Plasma adrenocorticotropic hormone (ACTH), beta-endorphin and alpha-melanocyte stimulating hormone were elevated after acoustic startle reflex testing in both WT and (Plg-/-) mice. However, in the Plg-/- mice, beta-endorphin values were 43, 35, and 45% lower in the plasma, pituitary, and whole brain, respectively, compared with the WT mice. Plasmin readily degraded precursor peptides, the 23-kDa precursor, beta-lipotropin, and ACTH, when presented as purified proteins or as the secretory products of mouse pituitary cells (AtT-20). The precursor peptide, 23 kDa, for beta-endorphin and alpha-melanocyte stimulating hormone was reduced in the pituitaries from the Plg-/- mice, and the mRNA for Plg was found in pituitaries from WT mice. Infusion of beta-endorphin and alpha-melanocyte stimulating hormone into the brain of Plg-/- mice increased acoustic startle reflex. The results of this study show that plasmin is involved in the processing of hormones derived from the pro-opiomelanocortin precursor in the intermediate pituitary. A deficiency of plasminogen reduces processing of beta-endorphin and alpha-melanocyte stimulating hormone, and interferes with normal brain function.