Merck
  • RELA/NEAT1/miR-302a-3p/RELA feedback loop modulates pancreatic ductal adenocarcinoma cell proliferation and migration.
RELA/NEAT1/miR-302a-3p/RELA feedback loop modulates pancreatic ductal adenocarcinoma cell proliferation and migration.
Journal of cellular physiology (2018-10-27)
Zhen Luo, Zhong-Jie Yi, Zheng-Lin Ou, Tong Han, Tao Wan, Yong-Chang Tang, Zhi-Chao Wang, Fei-Zhou Huang
ABSTRACT
Pancreatic ductal adenocarcinoma (PDAC) remains a challenging malignancy due to distant metastasis. RELA, a major component of the NF-κB pathway, could serve as an oncogene through activating proliferation or migration-related gene expression, including NEAT1, a well-known oncogenic long noncoding RNA. In the current study, the expression and function of RELA and NEAT1 in PDAC were examined. The potential upstream regulatory microRNAs of RELA were screened and verified for their correlation with RELA and NEAT1. The expression and function of the selected miR-302a-3p were evaluated. RELA and NEAT1 expression were upregulated in PDAC tissues, particularly in PDAC tissues with lymph node metastasis, and their expression correlated with clinical parameters. RELA overexpression promoted PDAC cell proliferation and migration, which could be partially attenuated by the NEAT1 knockdown. By binding to RELA, miR-302a-3p inhibited RELA expression, as well as PDAC cell proliferation and migration. RELA downstream NEAT1 expression was negatively regulated by miR-302a-3p; the suppressive effect of NEAT1 knockdown on PDAC cell proliferation and migration was partially attenuated by miR-302a-3p inhibition. Moreover, through direct binding, the expression of miR-302a-3p was also negatively regulated by NEAT1. The expression of miR-302a-3p was downregulated and negatively correlated with RELA or NEAT1 in tissue samples, indicating that rescuing miR-302a-3p expression may inhibit PDAC cell proliferation and migration through RELA/NEAT1. In summary, RELA, NEAT1, and miR-302a-3p form a feedback loop in PDAC to modulate PDAC cell proliferation and migration.