- Synthesis of coumarin-based, esterase-sensitive cyclic prodrugs of opioid peptides with enhanced membrane permeability and enzymatic stability.
Synthesis of coumarin-based, esterase-sensitive cyclic prodrugs of opioid peptides with enhanced membrane permeability and enzymatic stability.
Methods in molecular medicine (1999-01-01)
B Wang, D Shan, W Wang, H Zhang, O Gudmundsson, R T Borchardt
PMID21380892
ABSTRACT
With the discovery of an increasing number of biologically active peptides and peptide mimetics (1-3), there is a pressing need for the development of strategies to deliver these biologically active compounds to the desired site of action. The preceding two chapters have described two methods of making esterase-sensitive cyclic prodrugs of peptides. In this chapter, we wish to describe a third method of making esterase-sensitive cyclic prodrugs of peptides using DADLE, an opioid peptide (4-7), as an example.