Merck
  • Simultaneous Improvement of Efficiency and Lifetime of Quantum Dot Light-Emitting Diodes with a Bilayer Hole Injection Layer Consisting of PEDOT:PSS and Solution-Processed WO3.
Simultaneous Improvement of Efficiency and Lifetime of Quantum Dot Light-Emitting Diodes with a Bilayer Hole Injection Layer Consisting of PEDOT:PSS and Solution-Processed WO3.
ACS applied materials & interfaces (2018-06-27)
Ling Chen, Shujie Wang, Dongdong Li, Yan Fang, Huaibin Shen, Linsong Li, Zuliang Du
ABSTRACT
Even though chemically stable metal oxides (MOs), as substitutes for poly(3,4-ethylenedioxythiophene):polystyrene sulfonate (PEDOT:PSS), have been successfully adopted for improving device stability in solution-processed quantum dot light-emitting diodes (QLEDs), the efficiencies of QLEDs are at a relatively low level. In this work, a novel architecture of QLEDs has been introduced, in which inorganic/organic bilayer hole injection layers (HILs) were delicately designed by inserting an amorphous WO3 interlayer between PEDOT:PSS and the indium tin oxide anode. As a result, the efficiency and operational lifetime of QLEDs were improved simultaneously. The results show that the novel architecture QLEDs relative to conventional PEDOT:PSS-based QLEDs have an enhanced external quantum efficiency by a factor of 50%, increasing from 8.31 to 12.47%, meanwhile exhibit a relatively long operational lifetime (12 551 h) and high maximum brightness (>40 000 cd m-2) resulting from a better pathway for hole injection with staircase energy-level alignment of the HILs and reduction of surface roughness. Our results demonstrate that the novel architecture QLEDs using bilayer MO/PEDOT:PSS HILs can achieve long operational lifetime without sacrificing efficiency.