Merck
  • Identification of new interacting partners for atypical Rho GTPases: a SILAC-based approach.
Identification of new interacting partners for atypical Rho GTPases: a SILAC-based approach.
Methods in molecular biology (Clifton, N.J.) (2011-12-07)
Laura Montani, Damaris Bausch-Fluck, Ana Filipa Domingues, Bernd Wollscheid, João Bettencourt Relvas
ABSTRACT
In contrast to typical Rho GTPases the regulation of atypical Rho GTPases, such as the members of the RhoBTB subfamily, rarely depends on GEFs and/or GAPs. Instead, they are regulated at the level of their expression, by post-translational modifications, by their rate of degradation as well as through binding of diverse cell-specific interactors. Stable Isotope Labeling by Amino acids in Cell culture (SILAC) is a powerful cutting-edge mass-spectrometry-based technology allowing for protein-interaction studies in vitro with removal of false-positive identifications. In this chapter, we describe how the SILAC technology can be applied to the identification of new interacting partners for atypical - constitutively active - Rho GTPases, i.e. RhoBTB3.