Merck
  • Possible mechanisms of Leukoagglutinin induced apoptosis in human cells in vitro.
Possible mechanisms of Leukoagglutinin induced apoptosis in human cells in vitro.
Cell biology international (2016-09-16)
Tetiana Kochubei, Volodymyr Kitam, Oksana Maksymchuk, Oksana Piven, Lyubov Lukash
ABSTRACT
Leukoagglutinin is one of the phytohemagglutinin isolectins isolated from Phaseolus vulgaris. In our recent study, we showed that this lectin is able to influence the growth of human cancer cells in vitro. In addition, using the acridine orange and ethidium bromide staining, we found that leukoagglutinin can induce apoptosis. In order to understand the molecular mechanisms of induction of apoptosis, we performed computational modeling with subsequent experimental verification of theoretical data in vitro. We developed computational models of leukoagglutinin interaction with pro- (FasR and TNFR) and anti-apoptotic (IGF-1 and EGFR) receptors, and confirmed that leukoagglutinin may specifically interact with these receptors. Furthermore, we proved that leukoagglutinin can induce apoptosis in cancer (HEp-2) and non-cancer (4BL) cells, and observed that PHA-L is able to induce apoptosis through the up-regulation of Bax protein and activation of the effector caspase-3 and initiator caspase-8. However, these proteins have no effect on the Bcl-2 expression level.
Sigma-Aldrich
Anti-BAX antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution