Merck
  • Preparation of cyclodextrin chiral stationary phases by organic soluble catalytic 'click' chemistry.
Preparation of cyclodextrin chiral stationary phases by organic soluble catalytic 'click' chemistry.
Nature protocols (2011-07-02)
Yong Wang, Hui Chen, Yin Xiao, Cheong Hengq Ng, Ting Shan Oh, Timothy Thatt Yang Tan, Siu Choon Ng
ABSTRACT
We describe an effective and simple protocol that uses click chemistry to attach native β-cyclodextrin (β-CD) to silica particles, resulting in a chiral stationary phase (CCNCSP) that can be used for the enantioseparation of chiral drugs by high-performance liquid chromatography (HPLC). Starting from β-CD, the CCNCSP is prepared in several steps: (i) reaction of β-CD with 1-(p-toluenesulfonyl)-imidazole to afford mono-6-toluenesulfonyl-β-CD; (ii) azidolysis of mono-6-toluenesulfonyl-β-CD in dimethylformamide to give mono-6-azido-β-CD (N(3)-CD); (iii) reaction of cuprous iodide with triphenylphosphine to form an organic soluble catalyst CuI(PPh(3)); (iv) preparation of alkynyl-modified silica particles; and (v) click chemistry immobilization of N(3)-CD onto alkynyl-modified silica to afford the desired chiral stationary phase. Synthesis of the stationary phase and column packing takes ∼1 week.
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%