Merck
  • Therapeutic Response Monitoring with 89Zr-DFO-Pertuzumab in HER2-Positive and Trastuzumab-Resistant Breast Cancer Models.
Therapeutic Response Monitoring with 89Zr-DFO-Pertuzumab in HER2-Positive and Trastuzumab-Resistant Breast Cancer Models.
Pharmaceutics (2022-07-28)
Minwoo Kang, Jong Il Shin, Sangjin Han, Jung Young Kim, Jeonghoon Park, Kwang Il Kim, Joo Hyun Kang, Tae Sup Lee
ABSTRACT
Immuno-positron emission tomography (PET) has great potential to evaluate the target expression level and therapeutic response for targeted cancer therapy. Immuno-PET imaging with pertuzumab, due to specific recognition in different binding sites of HER2, could be useful for the determination of the therapeutic efficacy of HER2-targeted therapy, trastuzumab, and heat shock protein 90 (HSP90) inhibitor, in HER2-expressing breast cancer. The aim of this study is to evaluate the feasibility of monitoring therapeutic response with 89Zr-DFO-pertuzumab for the treatment of HER2-targeted therapeutics, trastuzumab, or the HSP90 inhibitor 17-DMAG, in trastuzumab-resistant JIMT-1 breast cancer models. We prepared an immuno-PET imaging agent using desferoxamine (DFO)-pertuzumab labeled with 89Zr and performed the biodistribution and PET imaging in breast cancer xenograft models for monitoring therapeutic response to HER2-targeted therapy. 89Zr-DFO-pertuzumab was successfully prepared and showed specific binding to HER2 in vitro and clearly visualized HER2 expressing JIMT-1 tumors. 89Zr-DFO-pertuzumab had prominent tumor uptake in HER2 expressing JIMT-1 tumors. JIMT-1 tumors showed trastuzumab-resistant and HSP90 inhibitor sensitive characterization. In immuno-PET imaging, isotype antibody-treated JIMT-1 tumors had similar uptake in trastuzumab-treated JIMT-1 tumors, but 17-DMAG-treated JIMT-1 tumors showed greatly reduced uptake compared to vehicle-treated tumors. Additionally, HER2 downregulation evaluated by immuno-PET imaging was verified by western blot analysis and immunofluorescence staining which resulted in a significant reduction in the tumor's HER2 level in 17-DMAG-treated JIMT-1 tumors. 89Zr-DFO-pertuzumab immuno-PET may be clinically translated to select pertinent patients for HER2-targeted therapy and to monitor the therapeutic response in HER2-positive cancer patients under various HER2-targeted therapeutics treatments.