Merck
  • Potent Anti-SARS-CoV-2 Activity by the Natural Product Gallinamide A and Analogues via Inhibition of Cathepsin L.
Potent Anti-SARS-CoV-2 Activity by the Natural Product Gallinamide A and Analogues via Inhibition of Cathepsin L.
Journal of medicinal chemistry (2021-11-04)
Anneliese S Ashhurst, Arthur H Tang, Pavla Fajtová, Michael C Yoon, Anupriya Aggarwal, Max J Bedding, Alexander Stoye, Laura Beretta, Dustin Pwee, Aleksandra Drelich, Danielle Skinner, Linfeng Li, Thomas D Meek, James H McKerrow, Vivian Hook, Chien-Te Tseng, Mark Larance, Stuart Turville, William H Gerwick, Anthony J O'Donoghue, Richard J Payne
ABSTRACT
Cathepsin L is a key host cysteine protease utilized by coronaviruses for cell entry and is a promising drug target for novel antivirals against SARS-CoV-2. The marine natural product gallinamide A and several synthetic analogues were identified as potent inhibitors of cathepsin L with IC50 values in the picomolar range. Lead molecules possessed selectivity over other cathepsins and alternative host proteases involved in viral entry. Gallinamide A directly interacted with cathepsin L in cells and, together with two lead analogues, potently inhibited SARS-CoV-2 infection in vitro, with EC50 values in the nanomolar range. Reduced antiviral activity was observed in cells overexpressing transmembrane protease, serine 2 (TMPRSS2); however, a synergistic improvement in antiviral activity was achieved when combined with a TMPRSS2 inhibitor. These data highlight the potential of cathepsin L as a COVID-19 drug target as well as the likely need to inhibit multiple routes of viral entry to achieve efficacy.