Merck
  • Effects of miR-211-3p/RHBDD1 axis on cell proliferation, cell cycle progression, and epithelial-mesenchymal transition in glioma.
Effects of miR-211-3p/RHBDD1 axis on cell proliferation, cell cycle progression, and epithelial-mesenchymal transition in glioma.
Folia neuropathologica (2022-08-12)
Lei Chen, Xiangyi Wang, Yude Zhu, Fuquan Liu, Laixing Liu, Ning Jiang
ABSTRACT
This study was designed to elucidate the relationship of miR-211-3p and rhomboid domain containing 1 (RHBDD1) in glioma. Here, we first observed that miR-211-3p directly targets the 3˘-UTR of RHBDD1 in glioma cells using dual-luciferase reporter assay, RNA immunoprecipitation (RIP) assay, reverse transcription-quantitative polymerase chain reaction (RT-qPCR), and Western blot analysis. Pearson's correlation analysis showed that miR-211-3p expression was negatively correlated with RHBDD1 expression in glioma tissues. CCK-8 assay, flow cytometry, and transwell assay were applied to assess cell proliferation, cell cycle distribution, migration, and invasion. The results showed that RHBDD1 knockdown inhibited cell proliferation, cell cycle G1/S transition, migration, and invasion in two glioma cell lines (U87 and LN-229). Knockdown of miR-211-3p obtained opposite results. Moreover, overexpression of RHBDD1 counteracted suppressive effects of miR-211-3p on glioma cells. Furthermore, decreased expression of CDK4, cyclin D1, N-cadherin, and vimentin as well as increased E-cadherin expression induced by miR-211-3p was reversed by RHBDD1 overexpression. Our results suggested that targeting miR-211-3p/RHBDD1 axis might be a novel effective therapeutic target for glioma treatment.