Merck
  • Magnesium Catalysis Mediated Tetrazoles in Desymmetrization Reaction of Aziridines.
Magnesium Catalysis Mediated Tetrazoles in Desymmetrization Reaction of Aziridines.
Organic letters (2017-05-31)
Dan Li, Kezhou Wang, Linqing Wang, Yuan Wang, Pengxin Wang, Xin Liu, Dongxu Yang, Rui Wang
ABSTRACT
A magnesium-catalyzed asymmetric ring-opening reaction of aziridines with substituted tetrazoles is reported. The current protocol proceeds smoothly and gives the corresponding desymmetrization products in high yields and good enantioselectivities. A new chiral ligand was synthesized from azetidine and (R)-BINOL and was employed in the current in situ generated magnesium catalyst. The Mg(II)-mediated desymmetrization reaction could be performed on gram scale under mild conditions and was transformed to chiral alkyl amines by a deprotection process.