- Visible-Light-Induced Photoaddition of N-Nitrosoalkylamines to Alkenes: One-Pot Tandem Approach to 1,2-Diamination of Alkenes from Secondary Amines.
Visible-Light-Induced Photoaddition of N-Nitrosoalkylamines to Alkenes: One-Pot Tandem Approach to 1,2-Diamination of Alkenes from Secondary Amines.
Organic letters (2021-04-02)
Dilip V Patil, Tengda Si, Hun Young Kim, Kyungsoo Oh
PMID33792333
ABSTRACT
The generation of aminium radical cation species from N-nitrosoamines is disclosed for the first time through visible-light excitation at 453 nm. The developed visible-light-promoted photoaddition reaction of N-nitrosoamines to alkenes was combined with the o-NQ-catalyzed aerobic oxidation protocol of amines to telescope the direct handling of harmful N-nitroso compounds, where the desired α-amino oxime derivatives were obtained in a one-pot tandem N-nitrosation and photoaddition sequence.