Merck
  • The effect of purified condensed tannins of forage plants from Botswana on the free-living stages of gastrointestinal nematode parasites of livestock.
The effect of purified condensed tannins of forage plants from Botswana on the free-living stages of gastrointestinal nematode parasites of livestock.
Veterinary parasitology (2013-08-10)
O Tibe, I A Sutherland, L Lesperance, D R K Harding
ABSTRACT
The effect of condensed tannins (CT) extracted from forage plants from Botswana on the free-living stages of a number of species of gastrointestinal nematode parasites derived from infected sheep were investigated using in vitro assays. Fresh samples of five different plants (Viscum rotundifolium, Viscum verrucosum, Tapinanthus oleifolius, Grewia flava and Ipomoea sinensis) were collected over two summers (February 2009 and 2010). Fractionation of each crude extract on a Sephadex LH-20 column yielded low molecular weight phenolics and CT-containing fractions. The effect of each purified CT fraction on parasites was evaluated using either egg hatch, larval development or larval migration inhibition assays. Three gastrointestinal nematode species (Haemonchus contortus, Trichostrongylus colubriformis and Teladorsagia circumcincta) derived from infected sheep were evaluated in the study. CT from V. rotundifolium and I. sinensis fractions from samples collected in 2009 and 2010 did not inhibit larval development. However, CT isolated from V. verrucosum, T. oleifolius and G. flava collected in 2009 completely inhibited the development of all parasite species. These CT fractions were more potent in inhibiting larval development of H. contortus than fractions from the same plant species collected in 2010. However, a slight effect on larval migration was observed with some CT extracts. The results suggest that CT extracts of some forage plants from Botswana have anti-parasitic properties in vitro, and that further research is required to determine any in vivo efficacy from feeding the plants to goats in a field situation.