Merck
  • Effects of metergoline on symptoms in women with premenstrual dysphoric disorder.
Effects of metergoline on symptoms in women with premenstrual dysphoric disorder.
The American journal of psychiatry (2002-11-02)
Catherine A Roca, Peter J Schmidt, Mark J Smith, Merry A Danaceau, Dennis L Murphy, David R Rubinow
ABSTRACT
The authors investigated the role of acute serotonergic modulation in the efficacy of selective serotonin reuptake inhibitors (SSRIs) in women with premenstrual dysphoric disorder. Patients with premenstrual dysphoric disorder (whose symptoms had remitted during treatment with fluoxetine) and a group of unmedicated healthy comparison women received the serotonin receptor antagonist metergoline as part of a double-blind, placebo-controlled crossover study. The patients with premenstrual dysphoric disorder experienced a return of symptoms 24 hours after treatment with metergoline but not diphenhydramine (active placebo). The comparison women experienced no changes in mood. These data support the role of altered serotonergic transmission in the efficacy of SSRI treatment for premenstrual dysphoric disorder.