Merck
Primary alterations during the development of hidradenitis suppurativa.
Journal of the European Academy of Dermatology and Venereology : JEADV (2021-11-02)
Z Dajnoki, O Somogyi, B Medgyesi, A Jenei, L Szabó, K Gáspár, Z Hendrik, P Gergely, D Imre, S Póliska, D Törőcsik, C C Zouboulis, E P Prens, A Kapitány, A Szegedi
ABSTRACT
Hidradenitis suppurativa (HS) is a chronic, inflammatory disease of the apocrine gland-rich (AGR) skin region. The initial steps of disease development are not fully understood, despite intense investigations into immune alterations in lesional HS skin. We aimed to systematically investigate the inflammatory molecules involved in three stages of HS pathogenesis, including healthy AGR, non-lesional HS and lesional HS skin, with the parallel application of multiple mRNA and protein-based methods. Immune cell counts (T cells, dendritic cells, macrophages), Th1/Th17-related molecules (IL-12B, TBX21, IFNG, TNFA, IL-17, IL10, IL-23A, TGFB1, RORC, CCL20), keratinocyte-related sensors (TLR2,4), mediators (S100A7, S100A8, S100A9, DEFB4B, LCN2, CAMP, CCL2) and pro-inflammatory molecules (IL1B, IL6, TNFA, IL-23A) were investigated in the three groups by RNASeq, RT-qPCR, immunohistochemistry and immunofluorescence. Epidermal changes were already detectable in non-lesional HS skin; the epidermal occurrence of antimicrobial peptides (AMPs), IL-1β, TNF-α and IL-23 was highly upregulated compared with healthy AGR skin. In lesional HS epidermis, TNF-α and IL-1β expression remained at high levels while AMPs and IL-23 increased even more compared with non-lesional skin. In the dermis of non-lesional HS skin, signs of inflammation were barely detectable (vs. AGR), while in the lesional dermis, the number of inflammatory cells and Th1/Th17-related mediators were significantly elevated. Our findings that non-lesional HS epidermal keratinocytes produce not only AMPs and IL-1β but also high levels of TNF-α and IL-23 confirm the driver role of keratinocytes in HS pathogenesis and highlight the possible role of keratinocytes in the transformation of non-inflammatory Th17 cells (of healthy AGR skin) into inflammatory cells (of HS) via the production of these mediators. The fact that epidermal TNF-α and IL-23 appear also in non-lesional HS seems to prove these cytokines as excellent therapeutic targets.