Merck
  • Osteocyte β1 integrin loss causes low bone mass and impairs bone mechanotransduction in mice.
Osteocyte β1 integrin loss causes low bone mass and impairs bone mechanotransduction in mice.
Journal of orthopaedic translation (2022-05-27)
Lei Qin, Tailin He, Dazhi Yang, Yishu Wang, Zhenjian Li, Qinnan Yan, Peijun Zhang, Zecai Chen, Sixiong Lin, Huanqing Gao, Qing Yao, Zhen Xu, Bin Tang, Weihong Yi, Guozhi Xiao
ABSTRACT
The key focal adhesion protein β1 integrin plays an essential role in early skeletal development. However, roles of β1 integrin expression in osteocytes during the regulation of bone homeostasis and mechanotransduction are incompletely understood. To study the in vivo function of osteocyte β1 integrin in bone, we utilized the 10-kb Dmp1 (Dentin matrix acidic phosphoprotein 1)-Cre to generate mice with β1 integrin deletion in this cell type. Micro-computerized tomography, bone histomorphometry and immunohistochemistry were performed to determine the effects of osteocyte β1 integrin loss on bone mass accrual and biomechanical properties. In vivo tibial loading model was applied to study the possible involvement of osteocyte β1 integrin in bone mechanotransduction. Loss of β1 integrin expression in osteocytes resulted in a severe low bone mass and impaired biomechanical properties in load-bearing long bones and spines, but not in non-weight-bearing calvariae, in mice. The loss of β1 integrin led to enlarged size of lacunar-canalicular system, abnormal cell morphology, and disorientated nuclei in osteocytes. Furthermore, β1 integrin loss caused shortening and disorientated collagen I fibers in long bones. Osteocyte β1 integrin loss did not impact the osteoclast activities, but significantly reduced the osteoblast bone formation rate and, in the meantime, enhanced the adipogenic differentiation of the bone marrow stromal cells in the bone microenvironment. In addition, tibial loading failed to accelerate the anabolic bone formation and improve collagen I fiber integrity in mutant mice. Our studies demonstrate an essential role of osteocyte β1 integrin in regulating bone homeostasis and mechanotransduction. The transnational potential of this article : This study reveals the regulatory roles of osteocyte β1 integrin in vivo for the maintenance of bone mass accrual, biomechanical properties, extracellular matrix integrity as well as bone mechanobiology, which defines β1 integrin a potential therapeutic target for skeletal diseases, such as osteoporosis.