Merck
  • Ammonium removal by erythrocyte-bioreactors based on glutamate dehydrogenase from Proteus sp. jointly with porcine heart alanine aminotransferase.
Ammonium removal by erythrocyte-bioreactors based on glutamate dehydrogenase from Proteus sp. jointly with porcine heart alanine aminotransferase.
Scientific reports (2022-04-02)
Daria V Borsakova, Larisa D Koleva, Evgeniy S Protasov, Fazoil I Ataullakhanov, Elena I Sinauridze
ABSTRACT
Excessive ammonium blood concentration causes many serious neurological complications. The medications currently used are not very effective. To remove ammonium from the blood, erythrocyte-bioreactors containing enzymes that processing ammonium have been proposed. The most promising bioreactor contained co-encapsulated glutamate dehydrogenase (GDH) and alanine aminotransferase (ALT). However, a low encapsulation of a commonly used bovine liver GDH (due to high aggregation), makes clinical use of such bioreactors impossible. In this study, new bioreactors containing ALT and non-aggregating GDH at higher loading were first produced using the flow dialysis method and the new bacterial GDH enzyme from Proteus sp. The efficacy of these erythrocyte-bioreactors and their properties (hemolysis, osmotic fragility, intracellular and extracellular activity of included enzymes, erythrocyte indices, and filterability) were studied and compared with native cells during 1-week storage. The ammonium removal rate in vitro by such erythrocyte-bioreactors increased linearly with an increase in encapsulated GDH activity. Alanine in vitro increased in accordance with ammonium consumption, which indicated the joint functioning of both included enzymes. Thus, novel bioreactors for ammonium removal containing GDH from Proteus sp. are promising for clinical use, since they have a more efficient GDH encapsulation and their properties are not inferior to previously obtained erythrocyte-bioreactors.