Merck
  • Lysosomes Dysfunction Causes Mitophagy Impairment in PBMCs of Sporadic ALS Patients.
Lysosomes Dysfunction Causes Mitophagy Impairment in PBMCs of Sporadic ALS Patients.
Cells (2022-04-24)
Matteo Bordoni, Orietta Pansarasa, Eveljn Scarian, Riccardo Cristofani, Roberta Leone, Valentina Fantini, Maria Garofalo, Luca Diamanti, Stefano Bernuzzi, Stella Gagliardi, Stephana Carelli, Angelo Poletti, Cristina Cereda
ABSTRACT
Mitochondria alterations are present in tissues derived from patients and animal models, but no data are available for peripheral blood mononuclear cells (PBMCs) of ALS patients. This work aims to investigate mitophagy in PBMCs of sporadic (sALS) patients and how this pathway can be tuned by using small molecules. We found the presence of morphologically atypical mitochondria by TEM and morphological abnormalities by MitoTracker™. We found a decreased number of healthy mitochondria in sALS PBMCs and an impairment of mitophagy with western blot and immunofluorescence. After rapamycin treatment, we found a higher increase in the LC3 marker in sALS PBMCs, while after NH4Cl treatment, we found a lower increase in the LC3 marker. Finally, mTOR-independent autophagy induction with trehalose resulted in a significant decrease in the lysosomes level sALS PBMCs. Our data suggest that the presence of morphologically altered mitochondria and an inefficient turnover of damaged mitochondria in PBMCs of sALS patients rely on the impairment of the mitophagy pathway. We also found that the induction of the mTOR-independent autophagy pathway leads to a decrease in lysosomes level, suggesting a more sensitivity of sALS PBMCs to trehalose. Such evidence suggests that trehalose could represent an effective treatment for ALS patients.