Merck
  • D1 and D2 dopamine receptor-mediated inhibition of activated normal T cell proliferation is lost in jurkat T leukemic cells.
D1 and D2 dopamine receptor-mediated inhibition of activated normal T cell proliferation is lost in jurkat T leukemic cells.
The Journal of biological chemistry (2010-07-02)
Biswarup Basu, Chandrani Sarkar, Debanjan Chakroborty, Subhalakshmi Ganguly, Saurav Shome, Partha Sarathi Dasgupta, Sujit Basu
ABSTRACT
Dopamine is a catecholamine neurotransmitter, which plays an important role in the regulation of T cell functions. In activated T cells from normal volunteers, stimulation of D(1) and D(2) dopamine receptors inhibit cell proliferation and cytokine secretion. However, there is no report yet regarding the regulatory role of D(1) and D(2) dopamine receptors in abnormally proliferating T cells. The present study investigates the expression and effect of activation of these dopamine receptors in Jurkat cells, a leukemic T cell line showing uncontrolled proliferation. Like normal human T cells, in Jurkat cells, D(1) and D(2) dopamine receptors are also expressed; however, unlike activated normal T cells, stimulation of these dopamine receptors in Jurkat cells fails to inhibit their T cell receptor-induced proliferation. This alteration is due to failure of D(1) dopamine receptor-mediated activation of cyclic AMP signaling and a missense mutation at the third cytoplasmic loop of D(2) dopamine receptors affecting inhibition of phosphorylation of ZAP-70, an important downstream protein transducing signal from the T cell receptor. These results help to understand the biology of abnormal proliferation of T cells in pathophysiological conditions where dopamine plays an important role.