Merck
  • Two Liberibacter Proteins Combine to Suppress Critical Innate Immune Defenses in Citrus.
Two Liberibacter Proteins Combine to Suppress Critical Innate Immune Defenses in Citrus.
Frontiers in plant science (2022-05-20)
Supratim Basu, Loan Huynh, Shujian Zhang, Roel Rabara, Hau Nguyen, Jeanette Velásquez Guzmán, Guixia Hao, Godfrey Miles, Qingchun Shi, Ed Stover, Goutam Gupta
ABSTRACT
We adopted a systems-based approach to determine the role of two Candidatus Liberibacter asiaticus (CLas) proteins, LasP 235 and Effector 3, in Huanglongbing (HLB) pathogenesis. While a published work suggests the involvement of these CLas proteins HLB pathogenesis, the exact structure-based mechanism of their action has not been elucidated. We conducted the following experiments to determine the structure-based mechanisms of action. First, we immunoprecipitated the interacting citrus protein partners of LasP 235 and Effector 3 from the healthy and CLas-infected Hamlin extracts and identified them by Liquid Chromatography with tandem mass spectrometry (LC-MS/MS). Second, we performed a split green fluorescent protein (GFP) assay in tobacco to validate that the interactions observed in vitro are also retained in planta. The notable in planta citrus targets of LasP 235 and Effector 3 include citrus innate immune proteins. Third, in vitro and in planta studies were performed to show that LasP 235 and Effector 3 interact with and inhibit the functions of multiple citrus proteins belonging to the innate immune pathways. These inhibitory interactions led to a high level of reactive oxygen species, blocking of bactericidal lipid transfer protein (LTP), and induction of premature programed cell death (PCD), all of which are beneficial to CLas lifecycle and HLB pathogenesis. Finally, we performed molecular dynamics simulations to visualize the interactions of LasP 235 and Effector 3, respectively, with LTP and Kunitz protease inhibitor. This led to the design of an LTP mimic, which sequestered and blocked LasP 235 and rescued the bactericidal activity of LTP thereby proving that LasP 235 , indeed, participates in HLB pathogenesis.