Merck
  • Pinoresinol diglucoside (PDG) attenuates cardiac hypertrophy via AKT/mTOR/NF-κB signaling in pressure overload-induced rats.
Pinoresinol diglucoside (PDG) attenuates cardiac hypertrophy via AKT/mTOR/NF-κB signaling in pressure overload-induced rats.
Journal of ethnopharmacology (2021-02-20)
Yusha Chen, Ruiyan Pan, Juanjuan Zhang, Tianming Liang, Juan Guo, Tai Sun, Xiaoyan Fu, Ling Wang, Lane Zhang
ABSTRACT
Pinoresinol diglucoside (PDG), the active compound extracted from Eucommia ulmoides, Styrax sp. and Forsythia suspensa, plays the roles in regulating hypertension, inflammation and oxidative stress. Considering that hypertension and inflammation has been proved to contribute to cardiac remodeling, we tested the effects of PDG on cardiac hypertrophy (CM). Male Sprague Dawley (SD) rats were used to construct hypertrophic rats by partial abdominal aortic constriction (AAC)-surgery. PDG solution (2 mg/ml) was used to treat AAC-induced rats by intraperitoneal injection at low dose (L-PDG, 2.5 mg/kg per day), medium dose (M-PDG, 5 mg/kg per day), and high dose (H-PDG, 7.5 mg/kg per day) for 3 weeks post AAC-surgery. CM was evaluated by the ratio of left ventricular weight to body weight ratio (LVW/BW), left ventricular wall thickness by H&E staining, and collagen content deposit by Masson's staining. Further, isoproterenol (ISO) and phenylephrine (PE) were used to produce cellular models of CM in neonatal rat ventricular cardiomyocytes (NRVMs). PDG pre-treated NRVMs 2 h at low dose (L-PDG, 2.5 μg/ml), medium dose (M-PDG, 5 μg/ml), and high dose (H-PDG, 7.5 μg/ml) for 24 h with or without PE- and ISO-stimulation. CM was evaluated by the expressions of hypertrophic biomarkers. Next, the hypertrophic biomarkers and pro-inflammatory cytokines were measured using quantitative real-time PCR (qRT-PCR), the expressions of protein kinase B (AKT)/mammalian target of rapamycin (mTOR)/transcription factor nuclear factor-kappa B (NF-kB) signaling pathway were determined by Western blotting. PDG treatment prevented cardiac histomorphology damages, decreased upregulations of hypertrophic biomarkers, and prevented fibrosis and inflammation after pressure overload resulting from AAC-surgery. Consistently, PDG remarkably inhibited the changes of cardiomyocyte hypertrophic biomarkers and inflammatory responses in cellular models of CM. Interestingly, PDG administration inhibited the activation of AKT/mTOR/NF-kB signaling pathway both in vivo and in vitro. PDG prevents AAC-induced CM in vivo, PE- and ISO-induced CM in vitro. The AKT/mTOR/NF-kB signaling pathway could be the potential therapeutic target involved in the protection of PDG. These findings provide novel evidence that PDG might be a promising therapeutic strategy for CM.