Merck
  • Multimodal profiling of the transcriptional regulatory landscape of the developing mouse cortex identifies Neurog2 as a key epigenome remodeler.
Multimodal profiling of the transcriptional regulatory landscape of the developing mouse cortex identifies Neurog2 as a key epigenome remodeler.
Nature neuroscience (2022-02-09)
Florian Noack, Silvia Vangelisti, Gerald Raffl, Madalena Carido, Jeisimhan Diwakar, Faye Chong, Boyan Bonev
ABSTRACT
How multiple epigenetic layers and transcription factors (TFs) interact to facilitate brain development is largely unknown. Here, to systematically map the regulatory landscape of neural differentiation in the mouse neocortex, we profiled gene expression and chromatin accessibility in single cells and integrated these data with measurements of enhancer activity, DNA methylation and three-dimensional genome architecture in purified cell populations. This allowed us to identify thousands of new enhancers, their predicted target genes and the temporal relationships between enhancer activation, epigenome remodeling and gene expression. We characterize specific neuronal transcription factors associated with extensive and frequently coordinated changes across multiple epigenetic modalities. In addition, we functionally demonstrate a new role for Neurog2 in directly mediating enhancer activity, DNA demethylation, increasing chromatin accessibility and facilitating chromatin looping in vivo. Our work provides a global view of the gene regulatory logic of lineage specification in the cerebral cortex.