Merck
  • Diagnostic value of glypican-3, arginase-1 and hepatocyte paraffin antigen -1 in differentiating hepatocellular carcinoma from intrahepatic cholangiocarcinoma.
Diagnostic value of glypican-3, arginase-1 and hepatocyte paraffin antigen -1 in differentiating hepatocellular carcinoma from intrahepatic cholangiocarcinoma.
Translational cancer research (2020-01-01)
Congrong Wang, Xiangyang Shao, Xuanyu Zhang, Chunmei Xie, Juanping Yu, Xiao Xu, Jian Yang, Yu Li, Weiwen Xu
ABSTRACT
The aim of the present study was to investigate the diagnostic value of glypican-3 (GPC3), arginase-1 (Arg-1), and hepatocyte paraffin antigen 1 (HepPar-1) in differentiating hepatocellular carcinoma (HCC) from intrahepatic cholangiocarcinoma (ICC). The expression of GPC3, HepPar-1 and Arg-1 were measured by immunohistochemistry in 47 cases of HCC, 29 cases of ICC and their paracancerous tissues. A high expression of GPC3, Arg-1 and HepPar-1 was observed in HCC tissues (68.09%, 76.60% and 78.72%, respectively; P>0.0125) while it was lower in ICC tissues (6.90%, 6.90% and 13.79%, respectively; P>0.0125). With regard to specificity, GPC3 performed better than Arg-1 and HepPar-1 (97.37% vs. 1.32% and 2.63%, respectively; P<0.05). The positive rate in poorly differentiated HCC for either GPC3, Arg-1 or HepPar-1 was lower than that in well- and moderately differentiated HCC. The majority of positive samples for GPC3 and Arg-1 were grade 2+ in well-, moderately- or poorly-differentiated HCC. Combined detection of GPC3, Arg-1 and HepPar-1 could increase the sensitivity up to 89.36% and the specificity to 100.00%, comparing with any single biomarker (P<0.05). GPC3, Arg-1 and HepPar-1 were all useful biomarkers in differentiating HCC from ICC. The combination models could improve the diagnosis value of HCC and help differentiating HCC from ICC.