Merck
  • Recognition of human proinsulin leader sequence by class I-restricted T-cells in HLA-A*0201 transgenic mice and in human type 1 diabetes.
Recognition of human proinsulin leader sequence by class I-restricted T-cells in HLA-A*0201 transgenic mice and in human type 1 diabetes.
Diabetes (2008-11-18)
Andréa Toma, Taghrid Laïka, Samy Haddouk, Sandrine Luce, Jean-Paul Briand, Luc Camoin, Francine Connan, Marion Lambert, Sophie Caillat-Zucman, Jean-Claude Carel, Sylviane Muller, Jeannine Choppin, François Lemonnier, Christian Boitard
ABSTRACT
A restricted region of proinsulin located in the B chain and adjacent region of C-peptide has been shown to contain numerous candidate epitopes recognized by CD8(+) T-cells. Our objective is to characterize HLA class I-restricted epitopes located within the preproinsulin leader sequence. Seven 8- to 11-mer preproinsulin peptides carrying anchoring residues for HLA-A1, -A2, -A24, and -B8 were selected from databases. HLA-A2-restricted peptides were tested for immunogenicity in transgenic mice expressing a chimeric HLA-A*0201/beta2-microglobulin molecule. The peptides were studied for binding to purified HLA class I molecules, selected for carrying COOH-terminal residues generated by proteasome digestion in vitro and tested for recognition by human lymphocytes using an ex vivo interferon-gamma (IFN-gamma) ELISpot assay. Five HLA-A2-restricted peptides were immunogenic in transgenic mice. Murine T-cell clones specific for these peptides were cytotoxic against cells transfected with the preproinsulin gene. They were recognized by peripheral blood mononuclear cells (PBMCs) from 17 of 21 HLA-A2 type 1 diabetic patients. PBMCs from 25 of 38 HLA-A1, -A2, -A24, or -B8 patients produced IFN-gamma in response to six preproinsulin peptides covering residues 2-25 within the preproinsulin region. In most patients, the response was against several class I-restricted peptides. T-cells recognizing preproinsulin peptide were characterized as CD8(+) T-cells by staining with peptide/HLA-A2 tetramers. We defined class I-restricted epitopes located within the leader sequence of human preproinsulin through in vivo (transgenic mice) and ex vivo (diabetic patients) assays, illustrating the possible role of preproinsulin-specific CD8(+) T-cells in human type 1 diabetes.