Merck
  • Enantioselective analysis of chiral anteiso fatty acids in the polar and neutral lipids of food.
Enantioselective analysis of chiral anteiso fatty acids in the polar and neutral lipids of food.
Lipids (2010-03-12)
Simone Hauff, Georg Hottinger, Walter Vetter
ABSTRACT
Anteiso fatty acids (aFA) are substituted with a methyl group on the antepenultimate carbon of the straight acyl chain. This feature leads to a stereogenic center. The 12-methyltetradecanoic acid (a15:0) and the 14-methylhexadecanoic acid (a17:0) are the most common aFA found in food, although they occur only in very small quantities. In this study we used gas chromatography in combination with a chiral stationary phase to determine the enantiomeric distribution of both a15:0 and a17:0 in the neutral and polar lipids of aquatic food samples and cheese. The best suited column was selected out of four custom-made combinations of heptakis(6-O-tert-butyldimethylsilyl-2,3-di-O-methyl)-beta-cyclodextrin (beta-TBDM) with different amount and polarity of an achiral polysiloxane. After separation of polar and neutral lipids of the food samples by solid phase extraction, fatty acid methyl esters were prepared and the fatty acid methyl esters were fractionated by reversed phase high performance liquid chromatography. Measurements of fractions high in aFA by enantioselective GC/MS in the selected ion monitoring mode verified the dominance of the (S)-enantiomers of a15:0 and a17:0 in both lipid fractions. However (R)-enantiomers were detectable in all samples. The relative proportion of the (R)-enantiomers was up to fivefold higher in the polar lipids than in the neutral lipids. The higher proportions in the polar lipids indicate that microorganisms might be involved in the formation of (R)-aFA.