Merck
  • Cross-reactivity of a pathogenic autoantibody to a tumor antigen in GABAA receptor encephalitis.
Cross-reactivity of a pathogenic autoantibody to a tumor antigen in GABAA receptor encephalitis.
Proceedings of the National Academy of Sciences of the United States of America (2021-02-24)
Simone M Brändle, Manuela Cerina, Susanne Weber, Kathrin Held, Amélie F Menke, Carmen Alcalá, David Gebert, Alexander M Herrmann, Hannah Pellkofer, Lisa Ann Gerdes, Stefan Bittner, Frank Leypoldt, Bianca Teegen, Lars Komorowski, Tania Kümpfel, Reinhard Hohlfeld, Sven G Meuth, Bonaventura Casanova, Nico Melzer, Eduardo Beltrán, Klaus Dornmair
ABSTRACT
Encephalitis associated with antibodies against the neuronal gamma-aminobutyric acid A receptor (GABAA-R) is a rare form of autoimmune encephalitis. The pathogenesis is still unknown but autoimmune mechanisms were surmised. Here we identified a strongly expanded B cell clone in the cerebrospinal fluid of a patient with GABAA-R encephalitis. We expressed the antibody produced by it and showed by enzyme-linked immunosorbent assay (ELISA) and immunohistochemistry that it recognizes the GABAA-R. Patch-clamp recordings revealed that it tones down inhibitory synaptic transmission and causes increased excitability of hippocampal CA1 pyramidal neurons. Thus, the antibody likely contributed to clinical disease symptoms. Hybridization to a protein array revealed the cross-reactive protein LIM-domain-only protein 5 (LMO5), which is related to cell-cycle regulation and tumor growth. We confirmed LMO5 recognition by immunoprecipitation and ELISA and showed that cerebrospinal fluid samples from two other patients with GABAA-R encephalitis also recognized LMO5. This suggests that cross-reactivity between GABAA-R and LMO5 is frequent in GABAA-R encephalitis and supports the hypothesis of a paraneoplastic etiology.
Sigma-Aldrich
Anti-GABA A Receptor α1 chain Antibody, NT, clone BD24, clone BD24, Chemicon®, from mouse